• USA Home
  • I8139 - L-myo-Inositol 1,4,5-tris­phosphate potassium salt

EMAIL THIS PAGE TO A FRIEND
I8139 Sigma

L-myo-Inositol 1,4,5-tris­phosphate potassium salt

≥98% (TLC)

  • CAS Number 129828-71-3

  • Empirical Formula (Hill Notation) C6H15O15P3

  • Molecular Weight 420.10

  •  PubChem Substance ID 24896133

Purchase

Properties

Related Categories Biochemicals and Reagents, Carbohydrates, Carbohydrates A to Z, Carbohydrates H-L, Monosaccharide More...
assay   ≥98% (TLC)
storage temp.   −20°C

Description

Other Notes

Enantiomer of the naturally occurring D-myo-inositol 1,4,5-trisphosphate. Used to study the stereoselectivity of phosphoinositide enzymes.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?