• USA Home
  • I8154 - InstaStain Ethidium Bromide Gel Staining Paper
EMAIL THIS PAGE TO A FRIEND
I8154 Sigma
InstaStain Ethidium Bromide Gel Staining Paper
size 14 cm × 14 cm
Purchase
Properties
size   14 cm × 14 cm
Description
General description
For staining nucleic acid electrophoresis gels. Ethidium bromide fixed on a special paper backing eliminates staining baths and reduces potential contact with ethidium bromide. Sensitivity is equivalent to staining with ethidium bromide solution. InstaStain is activated by microwaving the gel with a sheet of InstaStain on it, then allowing the gel to stand for 5 minutes followed by classical destaining. The solid format means much less waste than with traditional staining solutions. The 14 × 14 cm sheet can be cut for use with smaller gels.
Legal Information
InstaStain is a trademark of InstaStain, LLC.
InstaStain is a trademark of Edvotek
Price and Availability
Safety & Documentation
Safety Information
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?