• USA Home
  • I8378 - 5-Iodouridine 5′-monophosphate sodium salt

EMAIL THIS PAGE TO A FRIEND
I8378 Sigma

5-Iodouridine 5′-monophosphate sodium salt

~98%

Purchase

Properties

Related Categories Biochemicals and Reagents, Nucleosides, Nucleotides, Oligonucleotides, Nucleotide Analogs More...
InChI Key   DEGJKKOZBCELAN-UHFFFAOYSA-N
assay   ~98%
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?