• USA Home
  • I8508 - Imipramine hydrochloride

EMAIL THIS PAGE TO A FRIEND
I8508 Sigma

Imipramine hydrochloride

Sigma Reference Standard

Synonym: 10,11-Dihydro-N,N-dimethyl-5H-dibenz[b,f]azepine-5-propanamine hydrochloride, 5-[3-(Dimethylamino)propyl]-10,11-dihydro-5H-dibenz[b,f]azepine hydrochloride

Purchase

Properties

grade   Sigma Reference Standard
InChI Key   XZZXIYZZBJDEEP-UHFFFAOYSA-N
packaging   vial of 250 mg

Description

Packaging

Supplied in amber screw-cap vials

Biochem/physiol Actions

Tricyclic antidepressant; inhibits the serotonin and norepinephrine transporters with Kis of 7.7 nM and 67 nM, respectively. Has little effect on the dopamine transporter (Ki = 25 μM).

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
RTECS 
HO1925000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?