• USA Home
  • I8641 - Inhibin α-Subunit Fragment 1-32 porcine

EMAIL THIS PAGE TO A FRIEND
I8641 Sigma

Inhibin α-Subunit Fragment 1-32 porcine

≥90% (HPLC)

  • CAS Number 141983-93-9

  • Empirical Formula (Hill Notation) C163H248N48O43S

  • Molecular Weight 3600.07

  •  MDL number MFCD00151521

Purchase

Description

Amino Acid Sequence

Ser-Thr-Ala-Pro-Leu-Pro-Trp-Pro-Trp-Ser-Pro-Ala-Ala-Leu-Arg-Leu-Leu-Gln-Arg-Pro-Pro-Glu-Glu-Pro-Ala-Val-His-Ala-Asp-Cys-His-Arg

Application

An immunogenic fragment of inhibin.

Inhibin α-subunit fragment 1-32 porcine has been used in the ELISA (enzyme-linked immunosorbent assay) for detection of the inhibin antibody levels.

Biochem/physiol Actions

Inhibins are glycoproteins made of an α subunit and a β subunit. They suppress FSH (follicle stimulating hormone) secretion from the pituitary and also participate in control of gonadal functions. Inhibin α (INHA) is present in adrenal glands, pituitary gland, liver and placenta. It plays a significant role in male fertility.

Price and Availability


Amplified Detection

All labs need water
Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

HPLC Analysis of Inhibin α-Subunit Fragment 1-32 on Discovery® HS F5

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Inhibin α-Subunit Fragment 1-32 on Discovery® HS F5
Keywords: Chromatography, High performance liquid chromatography

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?