• USA Home
  • I8654 - Interferon-γ porcine

EMAIL THIS PAGE TO A FRIEND
I8654 Sigma

Interferon-γ porcine

>97% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder

Synonym: IFN-γ

Purchase

Properties

Related Categories Bioactives/Supplements, Cell Biology, Cell Signaling and Neuroscience, Cytokines and Growth Factors, Cytokines, Growth Factors and Hormones,
recombinant   expressed in E. coli
assay   >97% (SDS-PAGE)
form   lyophilized powder
mol wt   predicted mol wt ~17.3 kDa
impurities   endotoxin, tested
suitability   cell culture tested
Gene Information   pig ... IFNG(396991)

Description

Analysis Note

The biological activity is measured in an anti-viral assay using porcine PK-15 cells infected with EMC virus.

Biochem/physiol Actions

Cytokine with anti-viral, anti-proliferative, immunoregulatory, and proinflammatory activities.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline containing 2.5 mg bovine serum albumin.

Price and Availability


All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

related product

Product #

Description

Add to Cart

I8659 Anti-Interferon-γ antibody produced in goat, affinity isolated antibody, lyophilized powder

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?