EMAIL THIS PAGE TO A FRIEND
I8905 Sigma

L-Isoleucine

USP

Purchase

Properties

grade   USP

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

WGK Germany 
1

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

I7403 L-Isoleucine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?