• USA Home
  • I9008 - Indoxyl 1,3-diacetate

EMAIL THIS PAGE TO A FRIEND
I9008 Sigma

Indoxyl 1,3-diacetate

Synonym: 1,3-Diacetoxyindole

Purchase

Properties

Related Categories Building Blocks, C12, Chemical Synthesis, Heterocyclic Building Blocks, Indoles More...
InChI Key   DNVFBLDIZKYQPL-UHFFFAOYSA-N
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?