• USA Home
  • I9034 - Anti-Interleukin-1α propeptide antibody produced in sheep

EMAIL THIS PAGE TO A FRIEND
I9034 Sigma

Anti-Interleukin-1α propeptide antibody produced in sheep

affinity isolated antibody, lyophilized powder

Synonym: Anti-IL1, Anti-IL1A

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Cell Biology, Antibodies to Cytokines and Growth Factors, Antibodies to Interleukins (IL),
species reactivity   human
application(s)   flow cytometry: 25 μg/mL
  western blot: 0.1-0.2 μg/mL
clone   polyclonal
antibody form   affinity isolated antibody
form   lyophilized powder
mol wt   antigen mol wt ~14 kDa
storage temp.   −20°C
Gene Information   human ... IL1A(3552)
biological source   sheep
conjugate   unconjugated

Description

Immunogen

purified, E. coli-derived recombinant human interleukin-1α propeptide (aa 2-112).

General description

Interleukins are proinflammatory cytokines produced on cell injury or trauma. There are two closely related types of interleukins produced by the cell, IL-1α and IL-1β that are produced by the activation of NF-κ B transcription factor in response to bacterial infection or LPS. Both IL-1α and IL-1β exert their cellular effects by signalling through IL-1 receptor type 1. The IL-1α precursor produced initially is cleaved by calpain to produce a 16 kDa propeptide and 17 kDa mature protein. A variety of cells such as macrophages, epithelial cells, fibroblasts, keratinocytes, monocytes and dendritic cells secrete IL-1α. Both IL-1α and IL-1β are important in acquired immunity, inflammation, response to infection and production of matrix metalloproteinases (MMPs). The propeptide has nuclear localization sequence that enables it to shuttle to nucleus and activate transcription of target genes. The effects of IL-1α are mediated by activation of NF-κB, JNK and MAPK pathways that activated the genes in response to infection and inflammation.Overproduction of IL-1α has been implicated in inflammatory disorders like rheumatoid arthritis and periodontitis
Monoclonal Anti-Interleukin-1α propeptide recognizes human Interleukin-1α. This antibody shows less than 1% cross-reactivity with mature recombinant human Interleukin-1α.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline with 5% trehalose.

Application

Anti-interleukin 1α propeptide antibody is suitable for immunoblotting at a working concentration of 0.1-0.2 μg/ml. For flow cytometry, the recommended concentration is 25 μg/ml.

Price and Availability


Antibody Explorer

Explore with Confidence

All labs need water
Safety & Documentation

Safety Information

Personal Protective Equipment  Eyeshields, Gloves, type N95 (US), type P1 (EN143) respirator filter
RIDADR  NONH for all modes of transport
WGK Germany  1
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

I8284 Anti-Interleukin-1α antibody produced in goat, affinity isolated antibody, lyophilized powder
I8534 Anti-Interleukin-1α antibody produced in goat, affinity isolated antibody, lyophilized powder
I8909 Anti-Interleukin-1α antibody produced in goat, IgG fraction of antiserum, lyophilized powder
I7534 Monoclonal Anti-Interleukin-1α antibody produced in mouse, clone 4414, purified immunoglobulin, lyophilized powder
I7659 Monoclonal Anti-Interleukin-1α antibody produced in mouse, clone 3405, purified immunoglobulin, lyophilized powder
I7409 Monoclonal Anti-Interleukin-1α propeptide antibody produced in mouse, clone 409405, purified immunoglobulin, lyophilized powder
I5018 Anti-Interleukin-1α antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
I3392 Anti-Interleukin-1α antibody produced in goat, IgG fraction of antiserum, lyophilized powder

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?