• USA Home
  • I9156 - Monoclonal Anti-Itk antibody produced in mouse

EMAIL THIS PAGE TO A FRIEND
I9156 Sigma

Monoclonal Anti-Itk antibody produced in mouse

~1 mg/mL, clone 2F12, purified immunoglobulin, buffered aqueous solution

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Cell Biology, Antibodies for Kinase/Phosphatase Biology, IL2-Inducible T-Cell Kinase (Itk),
species reactivity   mouse, human
application(s)   immunoprecipitation: 2 μg using 1 mg protein lysate
  western blot: 1 μg/mL
clone   2F12, monoclonal
concentration   ~1 mg/mL
antibody form   purified immunoglobulin
form   buffered aqueous solution
isotype   IgG1
mol wt   antigen mol wt 72 kDa
shipped in   wet ice
storage temp.   −20°C
Gene Information   human ... ITK(3702)
mouse ... Itk(16428)
biological source   mouse
conjugate   unconjugated

Description

Immunogen

recombinant protein encoding the N-terminus of murine Itk.

Physical form

Solution in phosphate buffered saline pH. 7.4, containing 0.08% sodium azide.

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

Personal Protective Equipment  Eyeshields, Gloves
RIDADR  NONH for all modes of transport
WGK Germany  nwg
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?