• USA Home
  • I9253 - Invertase from baker's yeast (S. cerevisiae)

EMAIL THIS PAGE TO A FRIEND
I9253 Sigma

Invertase from baker's yeast (S. cerevisiae)

Grade V, Practical Grade, ≥30 units/mg solid, lactose and α-galactosidase (melibiase), essentially free

Synonym: β-D-Fructofuranosidase, β-D-Fructofuranoside fructohydrolase, Saccharase

Purchase

Description

Application

Used in the production of confectionary foods and artificial honey.

Biochem/physiol Actions

Invertase hydrolyzes sucrose into glucose and fructose yielding a colorless product, unlike acid hydrolysis which produces colored products.

Unit Definition

One unit will hydrolyze 1.0 μmole of sucrose to invert sugar per min at pH 4.5 at 55°C.

Price and Availability


Amplified Detection

Biomedical Applications
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles

Protocols

Enzymatic Assay of Invertase

This procedure applies to all products that have specification for the enzymatic activity of Invertase at Sigma-Aldrich Saint Louis.
Keywords: Biochemistry, Enzyme activity, Enzymology

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

I4504 Invertase from baker's yeast (S. cerevisiae), Grade VII, ≥300 units/mg solid

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?