Synonym: Kappa, Lambda Isotyping for Mouse Monoclonals, Mouse Monoclonal Antibody Isotyping Reagents
MDL number MFCD08705607
Popular Documents: Bulletin (PDF)
| Related Categories | Antibodies, Antibody Characterization and Analysis, Antibody Purification and Characterization, Biochemicals and Reagents, Isotyping Kits and Reagents, |
| storage temp. | 2-8°C |
Live Chat and Frequently Asked Questions
are available for this Product.
Determination of the subclass of a monoclonal antibody is helpful for characterization of the antibody, for choosing detection reagents, and for deciding on a purification scheme.
The IsoQuick strips, whether purchased in kits (ISOQ5 and ISOQ25) or separately (I9285, I9410, or I9535), are designed to detect and identify mouse isotypes in cell culture supernatant, ascites, or purified antibody. Optimum detection range is 0.1-4 μg/mL with a detection limit of 10 ng/mL. IsoQuick strips have been validated for use with mouse monoclonal antibodies and have not been validated for use with other host antibodies.
The strips come ready-to-use and provide results in 5 minutes or less. The IsoQuick strips can be used individually or in combination with the same prepared sample for customized results.
The IsoQuick Kappa/Lambda strips are designed to detect and identify mouse kappa or lambda isotype light chains.
IsoQuick is a trademark of EnviroLogix Inc.
Customers Also Viewed
sufficient for 5 applications
sufficient for 25 applications
IgG1, IgG2a, IgG2b, IgG3
IgA, IgM
sufficient for 1000 tests (clones) (by ELISA), sufficient for 40 tests (clones) (by immunodiffusion, ODD)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?