• USA Home
  • IDA5 - Iminodiacetic acid Agarose

EMAIL THIS PAGE TO A FRIEND
IDA5 Sigma

Iminodiacetic acid Agarose

Prepacked columns

Purchase

Properties

Related Categories Affinity Chromatography, Chelating Resins, Molecular Biology, Protein Chromatography, Proteomics More...
column volume   2.5 mL
storage temp.   2-8°C

Description

Application

Iminodiacetic acid-agarose is used in affinity chromatography, protein chromatography and chelating resins.

Price and Availability


Amplified Detection

Standard component

Description

Product #

Add to Cart

Iminodiacetic acid Agarose, saline suspension 5 column SDS I4758
Safety & Documentation

Safety Information

WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?