• USA Home
  • IMMH9 - S-100 Immunohistology Kit antibody produced in rabbit
EMAIL THIS PAGE TO A FRIEND
IMMH9 Sigma
S-100 Immunohistology Kit antibody produced in rabbit
Purchase
Description
Specificity
Contains primary antibody, biotinylated second antibody, ExtrAvidin-Peroxidase reagent, substrate buffer, H2O2 and 3-amino-9-ethylcarbazole.
Each kit contains reagents sufficient for a minimum of 30 slides.
Not for in vitro diagnostic use.
Price and Availability
Antibody Explorer
Explore with Confidence
Safety & Documentation
Safety Information
RIDADR 
UN 3316 9/PG 3
Documents
Certificate of Analysis
Protocols & Articles
Related Content
Antibody Explorer | Buy Primary & Secondary Antibodies
Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?