• USA Home
  • J105 - Anti-JunD antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
J105 Sigma

Anti-JunD antibody produced in rabbit

affinity isolated antibody, lyophilized powder

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Gene Regulation, Antibodies to Common Transcription Factors, Antibodies to Transcription Factors,
species reactivity   mouse
application(s)   western blot (chemiluminescent): 10 μg/mL using JunD expressed in induced/uninduced mouse cells
clone   polyclonal
antibody form   affinity isolated antibody
form   lyophilized powder
storage temp.   −20°C
Gene Information   mouse ... Jund(16478)
biological source   rabbit
conjugate   unconjugated

Description

Immunogen

synthetic peptide derived from amino acids 329-341 of the C-terminal end of mouse JunD.

Physical form

Lyophilized from phosphate buffered saline, containing 0.05% sodium azide.

Specificity

The antibody reacts specifically with JunD expressed in both uninduced and induced mouse fibroblasts (NIH 3T3 and L929) and also mouse pituitary corticotrophic line (AtT-20). It does not react with JunD in HeLa cells.

Reconstitution

Reconstitute with 100 μl deionized water.

Price and Availability

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?