• USA Home
  • J106 - Anti-JunB antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
J106 Sigma

Anti-JunB antibody produced in rabbit

affinity isolated antibody, lyophilized powder

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies to Common Transcription Factors, Antibodies to Transcription Factors, Cancer Research,
species reactivity   mouse
application(s)   western blot (chemiluminescent): 10 μg/mL
clone   polyclonal
antibody form   affinity isolated antibody
form   lyophilized powder
storage temp.   −20°C
Gene Information   mouse ... Junb(16477)
biological source   rabbit
conjugate   unconjugated

Description

Immunogen

synthetic peptide to corresponding to amino acids 45-61 of mouse JunB.

Physical form

Lyophilized from phosphate buffered saline containing 0.05% sodium azide.

Specificity

The antibody reacts specifically with JunB expressed in both uninduced and dexamethasone induced mouse fibroblast L929 cells. It does not react with JunB in HeLa cells.

Reconstitution

Reconstitute with 100 μl deionized water.

Price and Availability

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?