EMAIL THIS PAGE TO A FRIEND
J2125 Sigma

Janus Green B

certified by the BSC for supravital stain for blood, with neutral red

Purchase

Properties

clinical   for in vitro diagnostic use
suitability   certified by the BSC for supravital stain for blood, with neutral red

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
SG1633000

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?