EMAIL THIS PAGE TO A FRIEND
K1015 Sigma

K114

≥98% (HPLC), powder

Synonym: (trans,trans)-1-Bromo-2,5-bis-(4-hydroxy)styrylbenzene

  • CAS Number 872201-12-2

  • Empirical Formula (Hill Notation) C22H17BrO2

  • Molecular Weight 393.27

  •  MDL number MFCD11045294

Purchase

Properties

Related Categories Alzheimer′s Disease, Assays/Reagents for Alzheimer′s Disease Research, Bioactive Small Molecule Alphabetical Index, Bioactive Small Molecules, Cell Biology,
InChI Key   OXPHQQMZTXMEGO-RJTULKDBSA-N
assay   ≥98% (HPLC)
form   powder
color   white to beige
solubility   DMSO: soluble >10 mg/mL
storage temp.   2-8°C

Description

Biochem/physiol Actions

K114 is a fluorescent, amyloid-specific dye, an analogue of Congo Red and BSB that recognizes amyloid lesions and allows the quantitative monitoring of the formation of amyloid fibrils assembled from the Aβ peptide, α-synuclein, and tau. The affinity for fibrils ranges 20-30 nM. Unlike the other compounds K114 has minimal fluorescence in aqueous buffers, but fluoresces brightly in the presence of amyloid fibrils. K114 can be used to monitor in vitro amyloid fibril formation by fluorescence emission at 550 nm.

Features and Benefits

This compound is also offered as part of Sigma′s Library of Pharmacologically Active Compounds (LOPAC®1280), a biologically annotated collection of high-quality, ready-to-screen compounds. Click here to learn more.

Legal Information

LOPAC is a registered trademark of Sigma-Aldrich Co. LLC

Price and Availability


DISCOVER Bioactive Small Molecules

Making Headway

All labs need water
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?