• USA Home
  • K3501 - Anti-Human Kappa Light Chain (Bound and Free) antibody produced in goat

EMAIL THIS PAGE TO A FRIEND
K3501 Sigma

Anti-Human Kappa Light Chain (Bound and Free) antibody produced in goat

fractionated antiserum, lyophilized powder

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, HU-HZ, Human Kappa Secondary Antibodies and Conjugates, Primary Antibodies,
application(s)   quantitative precipitin assay: suitable
clone   polyclonal
antibody form   fractionated antiserum
form   lyophilized powder
storage temp.   2-8°C
Gene Information   human ... IGK@(50802)
biological source   goat
packaging   vial of 2 mL lyophilized antiserum
conjugate   unconjugated

Description

Physical form

Lyophilized from 0.01 M phosphate buffered saline, pH 7.2

Reconstitution

Reconstitute with 2 mL deionized water.

Price and Availability


Antibody Explorer

Explore with Confidence

Amplified Detection
Safety & Documentation

Safety Information

Personal Protective Equipment  Eyeshields, Gloves, type N95 (US), type P1 (EN143) respirator filter
RIDADR  NONH for all modes of transport
WGK Germany  3
Protocols & Articles

Articles

How to Choose a Secondary Antibody

The following information is provided to help you decide which secondary antibody may be best for your particular application.
Keywords: Amplification, Enzyme-linked immunosorbent assay, Flow cytometry, Immobilization, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western blot

Secondary Antibodies, Conjugates and Kits

Secondary antibodies are polyclonal or monoclonal antibodies that bind to primary antibodies or antibody fragments, such as the Fc or Fab regions. They are typically labeled with probes that make the...
Keywords: Absorption, Adsorption, Amplification, Bacterial conjugations, Cancer, Digestions, Electrophoresis, Enzyme-linked immunosorbent assay, Flow cytometry, Gel electrophoresis, Gene expression, Hormones, Immobilization, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunology, Immunoprecipitation, Infrared spectroscopy, Microbiology, Microscopy, Purification, Vitamins, Western blot

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Inflammation, Molecular biology, Phosphorylations, Purification, Western blot

Related Products

comparable product

Product #

Description

Add to Cart

K3502 Anti-Human Kappa Light Chain (Bound and Free) antibody produced in goat, affinity isolated antibody, lyophilized powder

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?