EMAIL THIS PAGE TO A FRIEND
K3764 Sigma

Kaliotoxin-1

recombinant, expressed in E. coli, ≥98% (HPLC), lyophilized powder

Synonym: 1KTX scorpion toxin

Purchase

Properties

Related Categories Bioactive Small Molecule Alphabetical Index, Bioactive Small Molecules, Cell Biology, Cell Signaling and Neuroscience, Ion Channels,
recombinant   expressed in E. coli
assay   ≥98% (HPLC)
form   lyophilized powder
shipped in   wet ice
storage temp.   −20°C

Description

Amino Acid Sequence

Gly-Val-Glu-Ile-Asn-Val-Lys-Cys-Ser-Gly-Ser-Pro-Gln-Cys-Leu-Lys-Pro-Cys-Lys-Asp-Ala-Gly-Met-Arg-Phe-Gly-Lys-Cys-Met-Asn-Arg-Lys-Cys-His-Cys-Thr-Pro

Biochem/physiol Actions

Blocker of Ca2+-activated and voltage-gated (KV1.1 and KV1.3) potassium channels

Preparation Note

Dissolve 1 μg in 0.24 ml of any conventional buffer for a stock solution of 1 μM.

Features and Benefits

This compound is featured on the Potassium Channels page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Price and Availability


Better Together

Discover your Target

Making Headway
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?