EMAIL THIS PAGE TO A FRIEND
K4387 Sigma

Katacine

95% (TLC), solid

Purchase

Properties

assay   95% (TLC)
form   neat
  solid
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
solubility   ethanol: 40%
  H2O: soluble
format   neat
storage temp.   2-8°C

Description

Biochem/physiol Actions

Anti-hypoxic

Caution

Protect from light.

Preparation Note

Mixture of polymers of proanthainanydines from the Polygonaceae family.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
2

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?