EMAIL THIS PAGE TO A FRIEND
K4505 Sigma

Kudzu root

lyophilized powder

Synonym: Radix puerariae

Purchase

Properties

Related Categories Biochemicals and Reagents, Protein Extracts, Proteins and Derivatives More...
form   lyophilized powder
storage temp.   2-8°C

Description

Preparation Note

Ethanol extract

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?