• USA Home
  • K5250 - 11-Ketoetiocholanolone glucuronide sodium salt

EMAIL THIS PAGE TO A FRIEND
K5250 Sigma

11-Ketoetiocholanolone glucuronide sodium salt

Synonym: 3α-Hydroxy-5β-androstane-11,17-dione-3α-ol 3-glucuronide, 5β-Androstane-11,17-dione-3α-ol 3-glucuronide

Purchase

Properties

storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?