• USA Home
  • L0145 - Laminarin azure from Laminaria digitata
EMAIL THIS PAGE TO A FRIEND
L0145 Sigma
Laminarin azure from Laminaria digitata
laminarinase substrate
Purchase
Description
Other Notes
Covalently linked with Remazol Brilliant Blue R
Preparation Note
Prepared by a modification of the procedure of Rinderknecht, H., et al., Experientia, 23, 805 (1967).
Substrates
Substrate for colorimetric determination of laminarinase.
Price and Availability
Biomedical Applications
Safety & Documentation
Safety Information
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Documents
Certificate of Analysis
Certificate of Origin
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?