L0169 Sigma

Anti-LRRK2 (C-terminal) antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution

DOWNLOAD MSDS (PDF)

Synonym: Anti-AURA17, Anti-Dardarin, Anti-Leucine-rich repeat protein kinase 2, Anti-PARK8, Anti-RIPK7, Anti-ROCO2

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Parkinson’s Disease Research, Antibodies in Array, Cell Biology,
species reactivity   human
application(s)   western blot: 1-2 μg/mL using HEK-293T cells expressing human LRRK2
clone   polyclonal
concentration   ~1.0 mg/mL
antibody form   affinity isolated antibody
form   buffered aqueous solution
mol wt   antigen mol wt ~240 kDa
shipped in   dry ice
storage temp.   −20°C
Gene Information   human ... LRRK2(120892)
mouse ... Lrrk2(66725)
rat ... (300160)

Description

Immunogen

synthetic peptide corresponding to amino acids 2512-2527 of human LRRK2 conjugated to KLH. The corresponding sequence is highly conserved (~80% identity) in mouse and rat LRRK2.

General description

LRRK2 is a μLtimeric kinase that also functions as a GTPase. LRRK2 activates a CaMKK-β/AMPK signaling pathway and subsequently mediates autophagy. Genetic alterations in LRRK2 cause late-onset Parkinson′s disease 1,2. Anti-LRRK2 (C-terminal) antibody is specific for human LRRK2 (approx. 240 kDa). Staining of the LRRK2 band by immunoblotting is specifically inhibited by the LRRK2 immunizing peptide.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Application

Anti-LRRK2 (C-terminal) antibody is suitable for use in western blot (1-2 μg/mL using HEK-293T cells expressing human LRRK2).

Price and Availability

Anti-Rabbit 2° Antibody Best Sellers


Prod. No.ConjugateSecondary Antibody Description
A0545HRPAnti-Rabbit IgG (whole molecule)-Peroxidase
A3687APAnti-Rabbit IgG (whole molecule)-Alkaline Phosphatase
F0382FITCAnti-Rabbit IgG (whole molecule)-FITC
B8895BiotinAnti-Rabbit IgG (whole molecule)-Biotin





Customers Also Viewed

Anti-LRRK2 (C-terminal region) antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution

Monoclonal Anti-LRRK2 antibody produced in mouse

~1.0 mg/mL, clone PROK57, purified immunoglobulin, buffered aqueous solution

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?