• USA Home
  • L0413 - Lipid Mixture 2, Chemically Defined

EMAIL THIS PAGE TO A FRIEND
L0413 Sigma

Lipid Mixture 2, Chemically Defined

cell culture tested, lyophilized powder

Purchase

Properties

Quality Level   GMP
sterility   aseptically processed
form   lyophilized powder
suitability   cell culture tested
storage temp.   2-8°C

Description

Application

Recommended for use in cell culture at 1 to 10 ml per liter of medium.

Other Notes

Contains non-animal derived fatty acids (2 μg/ml arachidonic and 10 μg/ml each linoleic, linolenic, myristic, oleic, palmitic and stearic), 0.22 mg/ml cholesterol from New Zealand sheep’s wool, 2.2 mg/ml Tween-80 and 70 μg/ml tocopherol acetate when reconsituted to the indicated volume in cell culture grade water (W 3500).

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
-

Documents

Certificate of Analysis

Protocols & Articles
Related Products

comparable product

Product #

Description

Add to Cart

L0288 Lipid Mixture 1, Chemically Defined, liquid, sterile-filtered, BioReagent, suitable for cell culture

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?