EMAIL THIS PAGE TO A FRIEND
L0505 Sigma

Lithium chloride

≥99.0% (titration)

Purchase

Properties

InChI Key   KWGKDLIKAYFUFQ-UHFFFAOYSA-M
assay   ≥99.0% (titration)
mp   605 °C(lit.)

Description

Other Notes

This product has been replaced by 213233-Sigma-Aldrich | Lithium chloride, ReagentPlus, 99% | LiCl

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
OJ5950000
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?