• USA Home
  • L0651 - Lectin from Lycopersicon esculentum (tomato)

EMAIL THIS PAGE TO A FRIEND
L0651 Sigma

Lectin from Lycopersicon esculentum (tomato)

biotin conjugate, lyophilized powder

Synonym: Lycopersicon esculentum agglutinin, LEA

Purchase

Properties

Related Categories Lycopersicon esculentum (Tomato), Glycan Recognizing Proteins, Glycobiology, Lectins, Molecular Biology,
form   lyophilized powder
composition   Lectin, ≥20%
extent of labeling   ≥3 mol biotin per mol lectin
conjugate   biotin conjugate
storage temp.   −20°C

Description

Analysis Note

Agglutination activity is expressed in μg/ml and is determined from serial dilutions in phosphate buffered saline, pH 7.3, of a 1 mg/ml solution. This activity is the lowest concentration to agglutinate a 2% suspension of human erythrocytes after 1 hour incubation at 25 °C.

Biochem/physiol Actions

LEA is not blood group specific, but has an affinity for N-acetyl-β-D-glucosamine oligomers. The lectin is a glycoprotein containing approx. equal amounts of protein and carbohydrate and is reported to inhibit the mitogenic activity of phytohemagglutinin from Phaseolus vulgaris. This lectin binds endothelial cells and is used to visualize the microvasculature.

Physical form

Contains NaCl

Price and Availability


Amplified Detection
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
nwg

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?