• USA Home
  • L0882 - L-Lysine Decarboxylase from Bacterium cadaveris

EMAIL THIS PAGE TO A FRIEND
L0882 Sigma

L-Lysine Decarboxylase from Bacterium cadaveris

Type V, partially purified powder, 1-3 units/mg protein (biuret)

Synonym: L-Lysine carboxy Lyase

Purchase

Properties

Related Categories 4.1.x.x C-C bonds, 4.x.x.x Lyases, Biochemicals and Reagents, Cell Biology, Enzyme Class Index,
type   Type V
form   partially purified powder
solubility   H2O: soluble
foreign activity   L-arginine decarboxylase, L-ornithine decarboxylase, L-histidine decarboxylase, L-glutamine decarboxylase and L-aspartic acid decarboxylase <0.2 unit/mg protein
storage temp.   −20°C

Description

Unit Definition

One unit will release 1.0 μmole of CO2 from L-lysine per min at pH 6.0 at 37°C.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?