• USA Home
  • L0883 - L-Lactic dehydrogenase

EMAIL THIS PAGE TO A FRIEND
L0883 Sigma

L-Lactic dehydrogenase

ammonium sulfate suspension, 400-900 units/mg protein

Purchase

Properties

form   ammonium sulfate suspension
storage temp.   2-8°C

Description

Analysis Note

Protein determined by biuret.

Physical form

Suspension in 3.2 M (NH4)2SO4, 5 mM potassium phosphate, pH 6.5

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?