EMAIL THIS PAGE TO A FRIEND
L117 Sigma

L-659,877

>95%, powder

  • CAS Number 125989-12-0

  • Empirical Formula (Hill Notation) C38H50N8O7S

  • Molecular Weight 762.92

  •  MDL number MFCD00076655

Purchase

Description

Amino Acid Sequence

Gln-Trp-Phe-Gly-Leu-Met

Biochem/physiol Actions

Potent and selective NK-2 tachykinin receptor antagonist.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Price and Availability


Detect Your Unseen Protein

All labs need water

Amicon Ultra Centrifugal Filters
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?