• USA Home
  • L1259 - Bandeiraea simplicifolia BS-II FITC labeled

EMAIL THIS PAGE TO A FRIEND
L1259 Sigma

Bandeiraea simplicifolia BS-II FITC labeled

lyophilized powder

Purchase

Properties

form   lyophilized powder
composition   Protein, ~95% E1%/280
storage temp.   −20°C

Description

Packaging

Package size based on protein content

Physical form

Lyophilized powder containing sodium citrate

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?