EMAIL THIS PAGE TO A FRIEND
L1268 Sigma

Linoleic acid

≥93% (GC)

Synonym: 9-cis,12-cis-Linoleic acid, cis-9,cis-12-Octadecadienoic acid, Telfairic acid

  • CAS Number 60-33-3

  • Linear Formula CH3(CH2)4CH=CHCH2CH=CH(CH2)7CO2H

  • Molecular Weight 280.45

  •  Beilstein Registry Number 1727101

  •  EC Number 200-470-9

  •  MDL number MFCD00064241

  •  PubChem Substance ID 24896275

  • Popular Documents:  FTNMR (PDF)  

Purchase

Properties

Related Categories Biochemicals and Reagents, Cell Biology, Fatty Acids and conjugates, Fatty Acyls, Functional Foods,
InChI Key   OYHQOLUKZRVURQ-HZJYTTRNSA-N
assay   ≥93% (GC)
form   liquid (clear)
refractive index   n20/D 1.466(lit.)
bp   229-230 °C/16 mmHg(lit.)
mp   −5 °C(lit.)
density   0.902 g/mL at 25 °C(lit.)
storage temp.   −20°C
Gene Information   human ... SOAT1(6646), SOAT2(8435)

Description

Biochem/physiol Actions

Linoleic acid increases cell proliferation and gene expression of PPARα and its target genes such as acyl-CoA oxidase in primary duck hepatocytes .

Price and Availability


Laboratory Gloves

The Future of Bioimaging

All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
RF9990000
Flash Point(F) 
>235.4 °F
Flash Point(C) 
>113 °C
Protocols & Articles

Articles

Omega-3 Fatty Acids and Heart Disease

The potential for the prevention and treatment of cardiovascular disease through increased dietary intake of omega-3 (w-3) fish oils is not a recent scientific discovery. A historically important stu...
BioFiles 2007, 2.2, 14.
Keywords: Anti-inflammatory agents, Biochemistry, Cardiovascular, Clinical, Degradations, Esterifications, Immunology, Inflammation, Lipid Metabolism, Metabolism, Metabolites, Methods, Nutrition, Oxidations, Transcription

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?