• USA Home
  • L1270 - Lectin from Glycine max (soybean)

EMAIL THIS PAGE TO A FRIEND
L1270 Sigma

Lectin from Glycine max (soybean)

Synonym: SBA, Soybean agglutinin

Purchase

Properties

storage temp.   −20°C

Description

Analysis Note

Agglutination activity is expressed in μg/ml and is determined from serial dilutions in phosphate buffered saline, pH 6.8, of a 1 mg per mL solution. This activity is the lowest concentration to agglutinate a 2% suspension of human blood group A erythrocytes after 1 hr incubation at 25 °C.

Biochem/physiol Actions

SBA is not blood group specific but has an affinity for N-acetyl-D-galactosamine.

Packaging

Package size based on protein content

Packaged in microcone vials for ease of reconstitution and recovery at microliter volumes.

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

comparable product

Product #

Description

Add to Cart

L2650 Lectin from Glycine max (soybean), peroxidase conjugate, lyophilized powder

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?