conjugate
peroxidase conjugate
form
lyophilized powder
peroxidase activity
10-30 units/mg protein
composition
Protein, ~90% biuret
storage temp.
−20°C
Looking for similar products? Visit Product Comparison Guide
Biochem/physiol Actions
DBA has anti-A1 human blood group specificity, useful for distinguishing between A1 and A2 blood types. DBA has strong anti-Cad activity, and has an affinity for terminal N-acetyl-α-D-galactosaminyl residues. Conjugates are prepared from affinity purified lectin.
Physical form
Contains sodium citrate
Preparation Note
Prepared from peroxidase type VI (P8375) using a modification of the method of O′Sullivan, which favors low molecular weight conjugates.
Other Notes
One unit will form 1 mg purpurogallin in 20 sec from pyrogallol at pH 6.0 at 20 °C.
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
A simple method for the preparation of enzyme-antibody conjugates.
M J O'Sullivan et al.
FEBS letters, 95(2), 311-313 (1978-11-15)
Laura Woods et al.
Nature communications, 8(1), 352-352 (2017-09-07)
Mammalian reproductive performance declines rapidly with advanced maternal age. This effect is largely attributed to the exponential increase in chromosome segregation errors in the oocyte with age. Yet many pregnancy complications and birth defects that become more frequent in older
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service