• USA Home
  • L138 - L-733,061 hydrochloride

EMAIL THIS PAGE TO A FRIEND
L138 Sigma

L-733,061 hydrochloride

solid

Synonym: (2R,3R)-3-[(3,5-bis(Trifluoromethyl)phenyl)methoxy]-2-phenylpiperidine hydrochloride

  • Empirical Formula (Hill Notation) C20H19F6NO · HCl

  • Molecular Weight 439.82

  •  MDL number MFCD06798982

  •  PubChem Substance ID 24896280

Purchase

Description

Biochem/physiol Actions

Weak NK-1 tachykinin receptor antagonist; less active enantiomer of L-733,060 hydrochloride.

Legal Information

Manufactured and sold under license from Merck, Sharp and Dohme Ltd.

Price and Availability


Amplified Detection

Making Headway

All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?