• USA Home
  • L144 - Mevinolin from Aspergillus sp.

EMAIL THIS PAGE TO A FRIEND
L144 Sigma

Mevinolin from Aspergillus sp.

powder

Synonym: 2-Methyl-1,2,3,7,8,8a-hexahydro-3,7-dimethyl-8-[2-(tetrahydro-4-hydroxy-6-oxo-2H-pyran-2-yl)ethyl]-1-naphthalenyl ester butanoic acid, 6-α-Methylcompactin, Lovastatin, Monacolin K

  • CAS Number 75330-75-5

  • Empirical Formula (Hill Notation) C24H36O5

  • Molecular Weight 404.54

  •  MDL number MFCD00072164

Purchase

Properties

InChI Key   PCZOHLXUXFIOCF-BXMDZJJMSA-N
form   powder
color   off-white
solubility   DMSO: >10 mg/mL
  ethanol: soluble16 mg/mL
  H2O: insoluble
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
RTECS 
EK7907000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

M2147 Mevinolin from Aspergillus sp., ≥98% (HPLC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?