• USA Home
  • L1538 - Anti-LIN-7 antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
L1538 Sigma

Anti-LIN-7 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym: Anti-MALS, Anti-Velis

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Cell Biology, Antibodies to Adhesion Molecules and Related Proteins, Antibodies to Cell and Organelle Proteins,
species reactivity   rat
application(s)   microarray: suitable
  western blot: 2.5 μg/mL using whole rat brain extract
clone   polyclonal
antibody form   affinity isolated antibody
form   buffered aqueous solution
mol wt   antigen mol wt 25-32 kDa (several bands)
shipped in   dry ice
storage temp.   −20°C
Gene Information   human ... LIN7A(8825)
rat ... Lin7a(85327)
biological source   rabbit
conjugate   unconjugated

Description

Immunogen

synthetic peptide corresponding to human LIN-7 (amino acids 191-205).

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 15 mM sodium azide.

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

WGK Germany  2
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?