• USA Home
  • L1637 - Luciferase, bacterial from Vibrio harveyi

EMAIL THIS PAGE TO A FRIEND
L1637 Sigma

Luciferase, bacterial from Vibrio harveyi

lyophilized powder

Purchase

Properties

form   lyophilized powder
composition   Protein, ~50% biuret
storage temp.   −20°C

Description

Features and Benefits

Partially purified, soluble extracts containing FMN-dependent luciferase and NADH- and NADPH-dependent FMN reductases. Produces light in a system containing FMN, NADH or NADPH, and n-decyl aldehyde.

Physical form

Lyophilized powder containing potassium phosphate buffer salts and stabilizer

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

comparable product

Product #

Description

Add to Cart

L8507 Luciferase from Vibrio fischeri (Photobacterium f), lyophilized powder

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?