• USA Home
  • L1894 - Lectin from Triticum vulgaris (wheat)

EMAIL THIS PAGE TO A FRIEND
L1894 Sigma

Lectin from Triticum vulgaris (wheat)

gold conjugate, 10 nm nominal (colloidal gold), aqueous glycerol suspension

Synonym: WGA, Wheat germ agglutinin

Purchase

Properties

form   aqueous glycerol suspension
concentration   ~5 A520 units/mL
nominal   10 nm (colloidal gold)
mean particle size   8-12 nm (monodisperse)
conjugate   gold conjugate
storage temp.   −20°C

Description

Biochem/physiol Actions

WGA is not blood group specific but has an affinity for N-acetyl-β-D-glucosaminyl residues and N-acetyl-β-D-glucosamine oligomers. WGA contains no protein-bound carbohydrate.

Preparation Note

Conjugates are prepared from affinity purified lectin.

Physical form

Suspension in approx. 50% glycerol containing 0.15 M NaCl, 0.01 M sodium phosphate, pH 7.2, 0.02% PEG 20, and 0.02% sodium azide

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?