• USA Home
  • L1913 - Anti-α-Latrotoxin antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
L1913 Sigma

Anti-α-Latrotoxin antibody produced in rabbit

IgG fraction of antiserum, lyophilized powder

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Neurobiology, Antibodies to Pharmacologicals, L-LL,
species reactivity   human, mouse, rat
application(s)   immunohistochemistry: 1:50
  western blot: 1:200
clone   polyclonal
antibody form   IgG fraction of antiserum
form   lyophilized powder
storage temp.   −20°C
Gene Information   human ... LPHN1(22859)
mouse ... Lphn1(330814)
rat ... Lphn1(65096)
biological source   rabbit
conjugate   unconjugated

Description

Immunogen

highly purified α-latrotoxin.

Physical form

Lyophilized from phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin, 5% sucrose, and 0.025% sodium azide.

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

WGK Germany  2
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Inflammation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?