• USA Home
  • L2014 - Lipoprotein, high density from human plasma

EMAIL THIS PAGE TO A FRIEND
L2014 Sigma

Lipoprotein, high density from human plasma

buffered aqueous solution, from human plasma

Synonym: α-Lipoprotein, HDL, High density lipoprotein

Purchase

Properties

biological source   from human plasma
sterility   0.22 μm filtered
form   buffered aqueous solution
shipped in   wet ice
storage temp.   2-8°C

Description

Caution

Freezing may cause denaturation.

Packaging

Package size based on protein content

Physical form

Solution in 0.15 M NaCl with 0.01% EDTA, pH 7.4

Price and Availability

Safety & Documentation

Safety Information

Protocols & Articles

Articles

Omega-3 Fatty Acids and Heart Disease

The potential for the prevention and treatment of cardiovascular disease through increased dietary intake of omega-3 (w-3) fish oils is not a recent scientific discovery. A historically important stu...
BioFiles 2007, 2.2, 14.
Keywords: Anti-inflammatory agents, Biochemistry, Cardiovascular, Clinical, Degradations, Esterifications, Immunology, Inflammation, Lipid Metabolism, Metabolism, Metabolites, Methods, Nutrition, Oxidations, Transcription

Related Products

comparable product

Product #

Description

Add to Cart

L8039 Lipoprotein, high density from human plasma, ≥95% (SDS-PAGE), solution

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?