• USA Home
  • L2130 - Lactoperoxidase from bovine milk

EMAIL THIS PAGE TO A FRIEND
L2130 Sigma

Lactoperoxidase from bovine milk

ammonium sulfate suspension, ≥50 units/mg protein

Synonym: Donor:hydrogen peroxide oxidoreductase, Peroxidase

Purchase

Properties

Related Categories 1.11.x.x Peroxide as receptor, 1.x.x.x Oxidoreductases, Biochemicals and Reagents, Cell Biology, Enzyme Class Index,
form   ammonium sulfate suspension
absorbance ratio   A412/280 nm 0.60-0.85
storage temp.   2-8°C
Gene Information   cow ... LPO(280844)

Description

Analysis Note

Protein determined by Lowry method.

Biochem/physiol Actions

Lactoperoxidase catalyzes the oxidation of iodide to iodine by hydrogen peroxide. This activity provides a gentle, specific alternative to chloramine T for the radioiodination of proteins and DNA.

Other Notes

Using 2,2′-azinobis(3-ethylbenzthiazoline-6-sulfonic acid) diammonium salt as substrate, two or three times the activity is observed.

Unit Definition

One unit will form 1.0 mg of purpurogallin from pyrogallol in 20 sec at pH 6.0 at 20 °C.

Physical form

Suspension in 3.2 M (NH4)2SO4 pH approx. 7.0

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.




All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles

Articles

Ammonium Sulfate Suspensions - Technical Note

Note that for an ammonium sulfate suspension, most of the enzyme will be in solid form. It is likely that only negligible amounts of enzyme will be in the ammonium sulfate solution.
Keywords: Enzyme activity

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

substrate

Product #

Description

Add to Cart

D9143 o-Dianisidine, peroxidase substrate
A0156 N-(4-Aminobutyl)-N-ethylisoluminol, ≥90%
A4685 Luminol sodium salt
A1888 2,2′-Azino-bis(3-ethylbenzothiazoline-6-sulfonic acid) diammonium salt, ≥98% (HPLC)
G5502 Guaiacol, oxidation indicator
A1661 N-(6-Aminohexyl)-N-ethylisoluminol
A3537 5-Aminosalicylic acid, ≥99%
A4382 4-Aminoantipyrine, reagent grade
A8264 4-Aminophthalhydrazide
A5754 3-Amino-9-ethylcarbazole, ≥95%, powder
A8511 Luminol, ≥97% (HPLC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?