• USA Home
  • L2170 - Anti-LRP1 (C-terminal) antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
L2170 Sigma

Anti-LRP1 (C-terminal) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym: Anti-α-2-Macroglobulin receptor, Anti-A2MR, Anti-APOER, Anti-APR, Anti-CD91, Anti-LRP, Anti-Low Density Lipoprotein receptor-related protein 1

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Alzheimer′s Disease, Antibodies for Neurobiology, Antibodies for Neurodegenerative Diseases Research,
species reactivity   rat, mouse, human
application(s)   indirect immunofluorescence: 1:500-1:1,000 using rat NRkcells
  western blot: 1:1,000-1:2,000 using whole extracts of mouseC2C12 cells
  western blot: 1:500-1:1,000 using whole extracts of human U87 cells
clone   polyclonal
antibody form   IgG fraction of antiserum
form   buffered aqueous solution
mol wt   antigen mol wt ~85 kDa
shipped in   dry ice
storage temp.   −20°C
Gene Information   human ... LRP1(4035)
mouse ... Lrp1(16971)
rat ... Lrp1(299858)
biological source   rabbit
conjugate   unconjugated

Description

Immunogen

synthetic peptide corresponding to amino acids 4532-4544 of human LRP1, conjugated to KLH. The corresponding sequence is identical in mouse, rat, dog, and monkey.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Enzyme-linked immunosorbent assay (1 paper)
Immunocytochemistry (1 paper)
Western Blotting (1 paper)

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

RIDADR  NONH for all modes of transport
Protocols & Articles

Articles

Alzheimer's Disease

Alzheimer's disease (AD) is the most common cause of dementia in the elderly and is characterized by gradual loss of cognitive functions. Hallmark pathohistological findings of AD include widespread ...
Carolyn L. Crankshaw
BioFiles v7 n2, 2011, 4–8
Keywords: Alzheimer Disease, Gene expression, Genetic, Genetics, Inflammation, Metabolism, Phosphorylations

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?