• USA Home
  • L2395 - D-Lactic Dehydrogenase from Leuconostoc mesenteroides

EMAIL THIS PAGE TO A FRIEND
L2395 Sigma

D-Lactic Dehydrogenase from Leuconostoc mesenteroides

ammonium sulfate suspension, 1,000-3,000 units/mg protein (biuret)

Synonym: (R)-Lactate:NAD+ oxidoreductase, D-LDH, Lactate

Purchase

Description

Packaging

5000 units in glass bottle

Unit Definition

One unit will reduce 1.0 μmole of pyruvate to D-lactate per min at pH 7.0 at 25 °C.

Physical form

Suspension in 3.2 M (NH4)2SO4, 0.1 M potassium phosphate, pH 7.0

Price and Availability


All labs need water

Biomedical Applications
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles

Articles

Ammonium Sulfate Suspensions - Technical Note

Note that for an ammonium sulfate suspension, most of the enzyme will be in solid form. It is likely that only negligible amounts of enzyme will be in the ammonium sulfate solution.
Keywords: Enzyme activity

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?