• USA Home
  • L2507 - Lectin from Arachis hypogaea (peanut)

EMAIL THIS PAGE TO A FRIEND
L2507 Sigma

Lectin from Arachis hypogaea (peanut)

agarose conjugate, saline suspension

Synonym: PNA, Peanut agglutinin

Purchase

Properties

Related Categories Arachis hypogaea (Peanut), Affinity Chromatography, Carbohydrate Binding Matrices, Glycan Recognizing Proteins, Glycobiology,
form   saline suspension
extent of labeling   2-4 mg per mL
matrix   cross-linked 4% beaded agarose
matrix activation   cyanogen bromide
matrix attachment   amino
matrix spacer   1 atom
capacity   ~1.5 mg/mL binding capacity (asialofetuin)
conjugate   agarose conjugate
storage temp.   2-8°C

Description

Application

Used for the purification of O-linked glycoproteins.

Biochem/physiol Actions

PNA does not agglutinate normal human erythrocytes, but strongly agglutinates neuraminidase treated erythrocytes. PNA has potent anti-T activity similar to the anti-T antibody in human sera. The lectin can be used to distinguish between human lymphocyte subsets.

Packaging

2 mL in glass bottle

Physical form

Suspension in 1.0 M NaCl containing 0.01% thimerosal

Price and Availability


Detect Your Unseen Protein

Amicon Pro Purification System
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?