• USA Home
  • L2536 - LY-310,762 hydrochloride

EMAIL THIS PAGE TO A FRIEND
L2536 Sigma

LY-310,762 hydrochloride

white powder

Synonym: 3,3-Dimethyl-1-{2-[4-(4-fluorobenzoyl)-1-piperidinyl]-1-ethyl}-1,3-dihydro-2H-indol-2-one hydrochloride

  • Empirical Formula (Hill Notation) C24H27O2N2F · HCl

  • Molecular Weight 430.94

  •  MDL number MFCD07772272

  •  PubChem Substance ID 24278114

Purchase

Description

Biochem/physiol Actions

Potent, selective 5-HT1D serotonin receptor antagonist.

Features and Benefits

This compound is also offered as part of Sigma′s Library of Pharmacologically Active Compounds (LOPAC®1280), a biologically annotated collection of high-quality, ready-to-screen compounds. Click here to learn more.

This compound was developed by Eli Lilly. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Legal Information

LOPAC is a registered trademark of Sigma-Aldrich Co. LLC

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?