• USA Home
  • L2638 - Lectin from Codium fragile (green marine algae)

EMAIL THIS PAGE TO A FRIEND
L2638 Sigma

Lectin from Codium fragile (green marine algae)

lyophilized powder

Purchase

Properties

Related Categories Codium fragile (Green marine algae), Glycan Recognizing Proteins, Glycobiology, Lectins, Molecular Biology,
form   lyophilized powder
potency   <2 μg per mL agglutination activity
impurities   salt, essentially free
storage temp.   −20°C

Description

Analysis Note

Agglutination activity is expressed in μg/ml and is determined from serial dilutions of a 1 mg/ml solution in saline (0.9% NaCl). This activity is the lowest concentration to agglutinate a 2% suspension of human blood group A erythrocytes after 1 hr incubation at 25 °C.

Biochem/physiol Actions

Codium fragile, subspecies tomentosides, agglutinin is not blood group specific, but has an affinity for N-acetyl-D-galatosamine.

Preparation Note

Purified by affinity chromatography.

Price and Availability


Amplified Detection
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?