• USA Home
  • L2776 - Lys-Gln-Ala-Gly-Asp-Val

EMAIL THIS PAGE TO A FRIEND
L2776 Sigma

Lys-Gln-Ala-Gly-Asp-Val

≥97% (HPLC)

Synonym: KQAGDV

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Other Peptides, Peptides and Proteins, Peptides for Cell Biology More...
InChI Key   AFAFFVKJJYBBTC-UHFFFAOYSA-N
assay   ≥97% (HPLC)
storage temp.   −20°C

Description

Amino Acid Sequence

Lys-Gln-Ala-Gly-Asp-Val

Packaging

1 mg in serum bottle

Biochem/physiol Actions

Lys-Gln-Ala-Gly-Asp-Val (KQAGDV) is a cell adhesion peptide used to study adhesion of platelets to neutrophils via the integrin glycoproteins gpIIb/IIIa.

Price and Availability


Detect Your Unseen Protein

All labs need water

Amicon Ultra Centrifugal Filters
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?