• USA Home
  • L3151 - L-Lactic Dehydrogenase from Bacillus stearothermophilus

EMAIL THIS PAGE TO A FRIEND
L3151 Sigma

L-Lactic Dehydrogenase from Bacillus stearothermophilus

recombinant, expressed in E. coli, lyophilized powder, ≥200 units/mg protein (Lowry)

Synonym: (S)-Lactate: NAD+ oxidoreductase, L-LDH, LAD, LD

Purchase

Description

Biochem/physiol Actions

Also catalyzes the oxidation of other L-2-hydroxymonocarboxylic acids.

Unit Definition

One unit will reduce 1.0 μmole of pyruvate to L-lactate per min at pH 6.0 at 30 °C.

Physical form

Lyophilized powder containing triethanolamine, buffer salts, EDTA and stabilizer

Price and Availability


All labs need water

Biomedical Applications
Safety & Documentation

Safety Information

WGK Germany 
3
Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Related Products

comparable product

Product #

Description

Add to Cart

L1254 L-Lactic Dehydrogenase from rabbit muscle, Type XI, lyophilized powder, 600-1,200 units/mg protein

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?